Tobramycin 95+%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol

45543-0010 45543-0050 45543-0250
TS45543-0010EA 67 USD
TS45543-0010 TS45543-0050 TS45543-0250
Tobramycin 95+%
Tobramycin
Formula: C₁₈H₃₇N₅O₉
Storage Temperature: Ambient
MDL Number: MFCD00077885
CAS Number: 32986-56-4

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤8 %
Titration with HClO4 ≥95.0 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR