N-Cbz-glycine(Z-Gly-OH) 98.5%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid

10835-0250 10835-1000 10835-5000
TS10835-0250EA 38 USD
TS10835-0250 TS10835-1000 TS10835-5000
N-Cbz-glycine(Z-Gly-OH) 98.5%
N-Cbz-glycine(Z-Gly-OH)
Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00002691
CAS Number: 1138-80-3
EINECS: 214-516-0

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 117 to 122 °C
HPLC ≥98.0 %
Titration with NaOH ≥98.0 %
Solubility light yellow solution
Titration with HClO4 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR