Erythromycin Ph. Eur.

Supplier: Thermo Scientific Chemicals

Synonyms: (-)-Erythromycin

34388-0050 34388-0250
TS34388-0050EA 79 USD
TS34388-0050 TS34388-0250
Erythromycin Ph. Eur.
Erythromycin
Formula: C₃₇H₆₇NO₁₃
MW: 733.95 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00084654
CAS Number: 114-07-8

Order Now

Specification Test Results

Appearance Colorless to white to light yellow Powder or crystals
Water ≤6.5 % (K.F.)
Sulfated ash ≤0.2 %
HPLC ≤5.0 % (erythromycin C)
Identification A: Passes test (Infrared spectrometry)
Related substances ≤7.0 % (HPLC) total impurities
Specific optical rotation -71° to -78° (20°C, 589 nm) (c=2, C2H5OH) (after 30 min, on anhydrous substance)
Thiocyanate (SCN) ≤0.3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR