Erythromycin 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (-)-Erythromycin

22733-0050 22733-0250 22733-1000
TS22733-0050EA 81 USD
TS22733-0050 TS22733-0250 TS22733-1000
Erythromycin 98%
Erythromycin
Formula: C₃₇H₆₇NO₁₃
MW: 733.95 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00084654
CAS Number: 114-07-8
EINECS: 204-040-1

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤6.5 % (K.F.)
Sulfated ash ≤0.2 %
HPLC ≥97.5 % (sum A,B and C)
Specific optical rotation -71° to -78° (20°C, 589 nm) (c=2, C2H5OH, 30 minutes) on dry substance

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR