Carbenicillin disodium salt 90%

Supplier: Thermo Scientific Chemicals
Danger

45536-0010 45536-0050
TS45536-0010EA 143 USD
TS45536-0010 TS45536-0050
Carbenicillin disodium salt 90%
Carbenicillin disodium salt
Formula: C₁₇H₁₆N₂O₆S·2Na
Boiling Pt: 738 °C
Melting Pt: >200 °C
Flash Pt: 400 °C
Storage Temperature: Freezer
MDL Number: MFCD00077683
CAS Number: 4800-94-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤6 %
Solubility (5 % in water) Clear to light yellow
HPLC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR