α-D-Glucose, anhydrous 99+%

Supplier: Thermo Scientific Chemicals

17008-0010 17008-0025 17008-0250
TS17008-0010EA 51 USD
TS17008-0010 TS17008-0025 TS17008-0250
α-D-Glucose, anhydrous 99+%
α-D-Glucose
Formula: C₆H₁₂O₆
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00063774
CAS Number: 492-62-6
EINECS: 207-757-8

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
HPLC ≥99.0 % (ELSD)
Appearance of solution (10g in 15ml water) Passes test
Related substances ≤0.15 % (Fructose)
Specific optical rotation +52.5° to +53.3° (20°C, 589 nm) (C=10,H2O/diluteNH3) (Anhydrous substance)
Soluble starch, sulfite ≤15 ppm
Dextrin Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR