Hydroxypropyl cellulose, M.W. ∼ 100.000

Supplier: Thermo Scientific Chemicals

18488-0010 18488-0051 18488-1000 18488-2500
TS18488-0010EA 594 USD
TS18488-0010 TS18488-0051 TS18488-1000 TS18488-2500
Hydroxypropyl cellulose, M.W. ∼ 100.000
Hydroxypropyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00132688
CAS Number: 9004-64-2

Order Now

Specification Test Results

Appearance White to cream Crystalline powder or granules
Infrared spectrum Conforms
Loss on drying ≤5 % (105°C)
Sulfated ash ≤0.5 %
Viscosity 75 to 150 mPa.s (5 % in water) (Brookfield settings: 30 rpm, spindle no.1)
pH 5 to 8.5 (1 % solution)
Arsenic (As) ≤3 ppm
Lead (Pb) ≤10 ppm
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR