2'-Deoxyuridine 99+%
Supplier: Thermo Scientific Chemicals
22661-0010
22661-0050
22661-2500
TS22661-0010EA
113
USD
TS22661-0010
TS22661-0050
TS22661-2500
2'-Deoxyuridine 99+%
2'-Deoxyuridine
Formula:
C₉H₁₂N₂O₅ MW: 228.20 g/mol Melting Pt: 163…167 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006527 CAS Number: 951-78-0 EINECS: 213-455-7 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % |
HPLC | ≥99.0 % |
Titration with NaOH | ≥99.0 % (On dry substance) |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...