N-Benzylglycine ethyl ester 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl 2-(benzylamino)acetate

10595-0100 10595-0500
TS10595-0100EA 38 USD
TS10595-0100 TS10595-0500
N-Benzylglycine ethyl ester 96%
N-Benzylglycine ethyl ester
Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Boiling Pt: 140…142 °C (10 mmHg)
Density: 1.03 g/cm³
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009174
CAS Number: 6436-90-4
EINECS: 229-218-6
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5045 to 1.5065 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR