Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl) 97%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Gly-OBzl·HCl, H-DL-Gly-OBzl hydrochloride, Benzyl glycinate hydrochloride, H-DL-Gly-OBzl.HCl

42854-0010 42854-0050 42854-0250
TS42854-0010EA 34 USD
TS42854-0010 TS42854-0050 TS42854-0250
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl) 97%
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl)
Formula: C₉H₁₂ClNO₂
MW: 201.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035442
CAS Number: 2462-31-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR consistent with structure
Iron (Fe) ≤0.0001 %
Heavy metals (as Pb) ≤1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR