3-Iodo-L-tyrosine 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Tyr(3-I)-OH, (S)-2-Amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
12244-0010
12244-0050
12244-0250
TS12244-0010EA
52
USD
TS12244-0010
TS12244-0050
TS12244-0250
3-Iodo-L-tyrosine 97%
3-Iodo-L-tyrosine
Formula:
C₉H₁₀INO₃ MW: 307.1 g/mol Melting Pt: 196…198 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002608 CAS Number: 70-78-0 EINECS: 200-744-8 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤4 % (K.F.) |
Solubility | (5 % in 1 N HCl) Clear colorless to light yellow |
HPLC | ≥96.0 % |
Specific optical rotation | -3° to -5° (20°C, 589 nm) (c=5 in 1 N HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...