Sarcosine methyl ester hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-Sar-OMe·HCl, N-Sar-OMe·HCl, Methyl N-methylglycinate hydrochloride
39750-0500
39750-2500
TS39750-0500EA
102
USD
TS39750-0500
TS39750-2500
Sarcosine methyl ester hydrochloride 97%
Sarcosine methyl ester hydrochloride
Formula:
C₄H₉NO₂·HCl MW: 139.58 g/mol Melting Pt: 110…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038876 CAS Number: 13515-93-0 |
Specification Test Results
Appearance | White to beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤1.0 % (1 g, 70°C) (to constant weight) |
Titration with HClO4 | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...