Sarcosine ethyl ester hydrochloride 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Sarcosine Ethyl Ester HCl, H-Sar-OEt·HCl, Ethyl 2-(methylamino)acetate hydrochloride

30559-0100 30559-0500
TS30559-0100EA 48 USD
TS30559-0100 TS30559-0500
Sarcosine ethyl ester hydrochloride 97%
Sarcosine ethyl ester hydrochloride
Formula: C₅H₁₁NO₂·HCl
MW: 153.61 g/mol
Melting Pt: 123…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00012506
CAS Number: 52605-49-9
EINECS: 258-037-5

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
Loss on drying ≤1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR