Procaine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
20731-1000
20731-5000
TS20731-1000EA
65
USD
TS20731-1000
TS20731-5000
Procaine hydrochloride 99%
Procaine hydrochloride
Formula:
C₁₃H₂₁ClN₂O₂ Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013000 CAS Number: 51-05-8 EINECS: 200-077-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Colorless to white to almost white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 154 to 158 °C |
Loss on drying | ≤0.5 % (1 g, 105 °C) |
Sulfated ash | ≤0.1 % |
pH | 5.0 to 6.5 (2 % w/v in water) |
Titration with NaOH | 98.5 to 101.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...