D-Panthenol 98+%
Supplier: Thermo Scientific Chemicals
23280-1000
23280-5000
TS23280-1000EA
82
USD
TS23280-1000
TS23280-5000
D-Panthenol 98+%
D-Panthenol
Formula:
C₉H₁₉NO₄ Boiling Pt: 118…120 °C (2.7Pa) Storage Temperature: Refrigerator |
MDL Number:
MFCD00065006 CAS Number: 81-13-0 EINECS: 201-327-3 |
Specification Test Results
Appearance | Clear colorless to light yellow Viscous liquid or solid |
Infrared spectrum | Conforms |
Refractive index | 1.4950 to 1.5020 (20°C, 589 nm) |
Assay | ≥98.0 % |
Water | ≤1.0 % (K.F.) |
Specific optical rotation | +29.0° to +32.0° (20°C, 589 nm) (c=5, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...