Melatonin 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide

12536-0010 12536-0050 12536-2500
TS12536-0010EA 82 USD
TS12536-0010 TS12536-0050 TS12536-2500
Melatonin 99%
Melatonin
Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
Boiling Pt: 512.8 °C
Melting Pt: 118 °C
Flash Pt: 264 °C
Storage Temperature: Ambient
MDL Number: MFCD00005655
CAS Number: 73-31-4
EINECS: 200-797-7

Order Now

Specification Test Results

Appearance White to cream to yellow Powder
Infrared spectrum Conforms
Melting point 116 to 120 °C
Loss on drying ≤0.5 % (1 g, 105 °C)
Heavy metals ≤20 ppm
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR