2,5-Piperazinedione 98%

Supplier: Thermo Scientific Chemicals

12008-0010 12008-0050 12008-0250 12008-1000
TS12008-0010EA 1194 USD
TS12008-0010 TS12008-0050 TS12008-0250 TS12008-1000
2,5-Piperazinedione 98%
2,5-Piperazinedione
Formula: C₄H₆N₂O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006009
CAS Number: 106-57-0
EINECS: 203-411-5

Order Now

Specification Test Results

Appearance White to light yellow to beige Crystalline powder
Infrared spectrum Conforms
Assay ≥97.5 % (ex N)
Loss on drying ≤0.5 % (105°C, 4 h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR