Palmitoleic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

37691-0050 37691-0010 37691-2500
TS37691-0050EA 717 USD
TS37691-0050 TS37691-0010 TS37691-2500
Palmitoleic acid 99%
Palmitoleic acid
Formula: C₁₆H₃₀O₂
Boiling Pt: 162 °C (.6 mmHg)
Melting Pt: .5 °C
Density: 0.894 g/cm³
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004437
CAS Number: 373-49-9
EINECS: 206-765-9

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4590 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR