N,N-Dimethylglycine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
N,N-Dimethylglycine HCl
20466-0050
20466-0250
20466-1000
TS20466-0050EA
58
USD
TS20466-0050
TS20466-0250
TS20466-1000
N,N-Dimethylglycine hydrochloride 99%
N,N-Dimethylglycine hydrochloride
Formula:
C₄H₁₀ClNO₂ MW: 139.58 g/mol Melting Pt: 188…191 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012610 CAS Number: 2491-06-7 EINECS: 219-648-2 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 188 to 198 °C |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...