Pyridoxine hydrochloride 98+%, extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI

15077-0500 15077-2500
TS15077-0500EA 99 USD
TS15077-0500 TS15077-2500
Pyridoxine hydrochloride 98+%, extra pure
Pyridoxine hydrochloride
Formula: C₈H₁₂ClNO₃
MW: 205.64 g/mol
Melting Pt: 214 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012807
CAS Number: 58-56-0
EINECS: 200-386-2
Merck Index: 13,08072

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Sulfated ash ≤0.1 %
Solubility Solution
pH 2.4 to 3.0 (5 % in water)
Titration with HClO4 98.0 to 102.0 % (On dry basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR