Vitamin A acetate 1.5 MIU/g in sunflower oil stabilized

Supplier: Thermo Scientific Chemicals
Danger

34632-1000 34632-5000
TS34632-1000EA 120 USD
TS34632-1000 TS34632-5000
Vitamin A acetate 1.5 MIU/g in sunflower oil stabilized
Vitamin A acetate
Formula: C₂₂H₃₂O₂
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00019413
CAS Number: 127-47-9
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Yellow to brown-yellow Partly crystalline when cooled
Assay 1.50 to 1.65 million IU/g (Vitamin A)
Identification (TLC): conforms
Stabilizer BHT
Related substances ≤0.14 (A370/A326)
Acid value ≤2.0 mg KOH/g
Peroxide value ≤15 meq/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR