Boc-Pro-OH 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid

27565-0250 27565-1000
TS27565-0250EA 79 USD
TS27565-0250 TS27565-1000
Boc-Pro-OH 99+%
Boc-Pro-OH
Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
Melting Pt: 133…136 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037324
CAS Number: 15761-39-4
EINECS: 239-848-3

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 133 to 136 °C
Water ≤0.5 % (K.F.)
HPLC ≥99.0 %
Specific optical rotation -59.5° to -61.5° (20 °C, 589 nm) (c=2, CH3COOH)
Titration with NaOH ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR