1-Aminocyclopropanecarboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

30142-0010 30142-2500
TS30142-0010EA 311 USD
TS30142-0010 TS30142-2500
1-Aminocyclopropanecarboxylic acid 99%
1-Aminocyclopropanecarboxylic acid
Formula: C₄H₇NO₂
MW: 101.11 g/mol
Melting Pt: 257 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00009944
CAS Number: 22059-21-8

Order Now

Specification Test Results

Appearance White to off-white Powder and/or granules
Infrared spectrum Conforms
Water ≤9 % (K.F.)
Titration with NaOH ≥98.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR