Adenine sulfate (synthetic) 98%

Supplier: Thermo Scientific Chemicals
Warning

16363-0250 16363-1000
TS16363-0250EA 136 USD
TS16363-0250 TS16363-1000
Adenine sulfate (synthetic) 98%
Adenine sulphate
Formula: C₅H₅N₅·½H₂SO₄
Melting Pt: 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00213655
CAS Number: 321-30-2
EINECS: 206-286-5

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (1 g, 105°C, 5 hrs) (vacuum)
Sulfated ash ≤0.1 % (800°C)
TLC Homogeneous
UV spectrophotometry molar absorptivity 13100 +/- 2%, on dry basis
UV transmittance ≥95 % (c=0.5, H2O, 430 nm) 1 cm cell
Ratio of Absorbances 0.34 to 0.38 (A280/A260)
Titration with NaOH ≥97.5 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR