Mycophenolic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

45938-0010 45938-0050 45938-0250
TS45938-0010EA 44 USD
TS45938-0010 TS45938-0050 TS45938-0250
Mycophenolic acid 98%
Mycophenolic acid
Formula: C₁₇H₂₀O₆
MW: 320.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036814
CAS Number: 24280-93-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 139 to 145 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR