L-allo-Threonine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-allo-Thr-OH, L(+)-allo-Threonine, Allo-Thr-OH

19854-1000 19854-2500
TS19854-1000EA 104 USD
TS19854-1000 TS19854-2500
L-allo-Threonine 99%
L-allo-Threonine
Formula: C₄H₉NO₃
MW: 119.12 g/mol
Melting Pt: 256 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064268
CAS Number: 28954-12-3
EINECS: 249-327-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Specific optical rotation +8° to +10° (20°C, 589 nm) (c=2, water)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR