L-Threonine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid

13893-0050 13893-0250 13893-1000 13893-5000
TS13893-0050EA 23 USD
TS13893-0050 TS13893-0250 TS13893-1000 TS13893-5000
L-Threonine 98%
L-Threonine
Formula: C₄H₉NO₃
Melting Pt: 255 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or glistening flakes
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 3 hrs)
Sulfated ash ≤0.1 %
Solubility (2.5 % in water) Clear colorless
Specific optical rotation -27.6° to -29.0° (20°C, 589 nm) (c=6, H2O)
Titration with HClO4 97.5 to 102.5 % (On dried substance)
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤300 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR