DL-Threonine 99.5%

Supplier: Thermo Scientific Chemicals

13894-0250 13894-1000 13894-5000
TS13894-0250EA 51 USD
TS13894-0250 TS13894-1000 TS13894-5000
DL-Threonine 99.5%
DL-Threonine
Formula: C₄H₉NO₃
MW: 119.12 g/mol
Melting Pt: 244 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00063722
CAS Number: 80-68-2
EINECS: 201-300-6

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.2 % (1 g, 105°C, 3 hrs)
Sulfated ash ≤0.1 %
Titration with HClO4 98.0 to 101.0 % (On dried substance)
Chloride (Cl) ≤200 ppm
Arsenic (As) ≤2 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids (TLC) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR