Cytochalasin B

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 7(S),20(R)-Dihydroxy-16(R)-methyl-10-phenyl-24-oxa[14]cytochalasa-6(12),13(E),21(E)-triene-1,23-dione

22809-0010 22809-0050 22809-0250
TS22809-0010EA 94 USD
TS22809-0010 TS22809-0050 TS22809-0250
Cytochalasin B
Cytochalasin B
Formula: C₂₉H₃₇NO₅
MW: 479.62 g/mol
Melting Pt: 218…223 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077704
CAS Number: 14930-96-2
EINECS: 239-000-2
UN: 1544
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to off-white Powder
HPLC ≥98 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR