Bicine 99+% for biochemistry
Supplier: Thermo Scientific Chemicals
32771-0250
32771-1000
32771-5000
TS32771-0250EA
44
USD
TS32771-0250
TS32771-1000
TS32771-5000
Bicine 99+% for biochemistry
Bicine
Formula:
C₆H₁₃NO₄ Melting Pt: 190…192 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004295 CAS Number: 150-25-4 EINECS: 205-755-1 |
Specification Test Results
Appearance | White Crystalline powder or crystals or needles |
Infrared spectrum | Conforms |
Water | ≤1 % (K.F.) |
Sulfated ash | ≤0.1 % |
UV | at 280 nm A: ≤0.05 |
Titration with NaOH | ≥99.0 % |
Iron (Fe) | ≤5 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...