Acacetin

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5,7-Dihydroxy-2-(4-methoxyphenyl)chromen-4-one, Linarisenin, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one, 4'-Methoxy-5,7-dihydroxyflavone, 5,7-Dihydroxy-4'-methoxyflavone, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4-benzopyrone, Linarigenin, Buddleoflavonol, 4'-Methoxyapigenin

46523-0010 46523-2500
TS46523-0010EA 169 USD
TS46523-0010 TS46523-2500
Acacetin
Acacetin
Formula: C₁₆H₁₂O₅
MW: 284.27 g/mol
Melting Pt: 260…265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016936
CAS Number: 480-44-4

Order Now

Specification Test Results

Appearance White to light yellow to light brown Powder or crystals
Infrared spectrum Conforms
Water ≤3 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR