CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulfonic acid) 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid
32774-0010
32774-0250
32774-1000
TS32774-0010EA
664
USD
TS32774-0010
TS32774-0250
TS32774-1000
CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulfonic acid) 99%
CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid)
Formula:
C₉H₁₉NO₄S MW: 237.32 g/mol Melting Pt: 263…271 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041778 CAS Number: 73463-39-5 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Water | ≤1 % (K.F.) |
Solubility | (0.1M in water) Clear colorless solution |
UV | at 280 nm A: ≤0.03 |
pH | 5 to 6.5 (1 % in water at 25°C) |
Titration with NaOH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...