(+)-Cinchonine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol
22781-0250
22781-1000
22781-5000
TS22781-0250EA
51
USD
TS22781-0250
TS22781-1000
TS22781-5000
(+)-Cinchonine 99%
(+)-Cinchonine
Formula:
C₁₉H₂₂N₂O MW: 294.4 g/mol Melting Pt: 255…265 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064372 CAS Number: 118-10-5 EINECS: 204-234-6 Merck Index: 13,02308 |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 255 to 265 °C |
Loss on drying | ≤1.0 % |
Sulfated ash | ≤0.1 % |
Specific optical rotation | +251° to +269° (20 °C, 589 nm) (c=2, 0.2M HCl) |
Titration with HClO4 | 98.5 to 101.5 % |
Other alkaloids | ≤3 % (HPLC) Quinine + hydroquinine |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...