Carboxymethyl cellulose sodium salt, M.W. ∼ 90000 (DS=0.7)

Supplier: Thermo Scientific Chemicals

Synonyms: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

33260-0010 33260-0050 33260-1000
TS33260-0010EA 137 USD
TS33260-0010 TS33260-0050 TS33260-1000
Carboxymethyl cellulose sodium salt, M.W. ∼ 90000 (DS=0.7)
Carboxymethyl cellulose sodium salt
Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS Number: 9004-32-4

Order Now

Specification Test Results

Appearance White to light yellow Granular powder
Infrared spectrum Conforms
Assay ≥99.5 %
Loss on drying ≤10 % (As packed) (3 to 5 g, 105°C, 2 h)
Heavy metals ≤20 ppm
Degree of substitution 0.65 to 0.90
Viscosity 50 to 100 mPa.s (2 % at 25°C) (Brookfield)
pH 6.5 to 8 (1 % solution)
Sodium chloride (NaCl) ≤0.25 %
Arsenic (As) ≤3 ppm
Cadmium (Cd) ≤1 ppm
Lead (Pb) ≤10 ppm
Mercury (Hg) ≤1 ppm
Impurity ≤0.4 % (Sodium glycolate)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR