Thiamine hydrochloride 98.5-101.5%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

14899-0100 14899-1000 14899-0010
TS14899-0100EA 52 USD
TS14899-0100 TS14899-1000 TS14899-0010
Thiamine hydrochloride 98.5-101.5%
Thiamine hydrochloride
Formula: C₁₂H₁₈Cl₂N₄OS
MW: 337.27 g/mol
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012780
CAS Number: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤5 % (K.F.)
Sulfated ash ≤0.1 %
Titration with HClO4 98.5 to 101.5 % (On anhydrous substance)
Sulfate (SO4) ≤300 ppm
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR