Alginic acid
Supplier: Thermo Scientific Chemicals
27058-0025
27058-1000
27058-5000
TS27058-0025EA
902
USD
TS27058-0025
TS27058-1000
TS27058-5000
Alginic acid
Alginic acid
Formula:
(C₆H₈O₆)n Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081309 CAS Number: 9005-32-7 EINECS: 232-680-1 |
Specification Test Results
Appearance | White to yellow to light beige Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤15 % (2 g, 105°C, 4 hrs) |
Sulfated ash | ≤8 % |
Titration excess NaOH | 19 to 25 % (Carboxyl groups) On dried substance |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...