Colchicine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine

22712-0010 22712-0050 22712-0250 22712-5000
TS22712-0010EA 120 USD
TS22712-0010 TS22712-0050 TS22712-0250 TS22712-5000
Colchicine 97%
Colchicine
Formula: C₂₂H₂₅NO₆
MW: 399.44 g/mol
Melting Pt: 150…160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00078484
CAS Number: 64-86-8
EINECS: 200-598-5
UN: 1544
ADR: 6.1,I
Merck Index: 12,02536

Order Now

Specification Test Results

Appearance Light yellow Powder
Infrared spectrum Conforms
Water ≤2 % (K.F.)
HPLC ≥96.0 % on anhydrous and solvent-free substance
Specific optical rotation -240° to -250° (20°C, 589 nm) (c=1, alcohol) (on anhydrous and solvent-free substance

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR