D-α-Phenylalanine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: D-Phenylalanine, H-D-Phe-OH, 3-Phenyl-D-alanine, 3-Phenyl-D(+)-alanine

17204-0250 17204-1000
TS17204-0250EA 98 USD
TS17204-0250 TS17204-1000
D-α-Phenylalanine 99+%
D-α-Phenylalanine
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 273…276 °C
Storage Temperature: Ambient
MDL Number: MFCD00004270
CAS Number: 673-06-3
EINECS: 211-603-5
Merck Index: 12,07425

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
Specific optical rotation +32° to +35° (20°C, 589 nm) (c=2, water)
Titration with HClO4 ≥99.0 %
Arsenic (As) ≤3 ppm
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR