DL-α-Phenylalanine 99%

Supplier: Thermo Scientific Chemicals

16457-0250 16457-1000 16457-5000
TS16457-0250EA 41 USD
TS16457-0250 TS16457-1000 TS16457-5000
DL-α-Phenylalanine 99%
DL-α-Phenylalanine
Formula: C₉H₁₁NO₂
Melting Pt: 266 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064225
CAS Number: 150-30-1
EINECS: 205-756-7
Merck Index: 13,07355

Order Now

Specification Test Results

Appearance White Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 %
Sulfated ash ≤0.3 %
Titration with HClO4 98.5 to 101.5 % (on dry substance)
Arsenic (As) ≤3 ppm
Lead (Pb) ≤10 ppm
Total nitrogen ≥9.0 % (on dry substance)
Amino nitrogen ≥3.0 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR