Oil of castor, meets analytical specification of Ph.Eur.

Supplier: Thermo Scientific Chemicals

Synonyms: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi

34885-0025 34885-5000
TS34885-0025EA 320 USD
TS34885-0025 TS34885-5000
Oil of castor, meets analytical specification of Ph.Eur.
Oil of castor
Boiling Pt: 229 °C (760 mmHg)
Melting Pt: –10 °C
Density: 0.957 g/cm³
Flash Pt: 229 °C
Storage Temperature: Ambient
MDL Number: MFCD00130746
CAS Number: 8001-79-4
EINECS: 232-293-8
Merck Index: 13,01908

Order Now

Specification Test Results

Appearance Clear colorless to slightly yellow Hygroscopic viscous liquid
Water ≤0.3 %
Identification A Passes test
Angle of optical rotation +3.5° to +6.0° (20°C, 589 nm) (neat)
Absorbance ≤0.7 (270 nm, 1 % in EtOH 96%)
Composition of fatty acids ≤1.0 % (Any other fatty acid)
Acid value ≤1.5 mg KOH/g
Hydroxyl value ≥160 mg KOH/g
Peroxide value ≤10.0 meq O2/kg
Unsaponifiable matter ≤0.8 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR