Methacrylamide 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

14920-0025 14920-2500 14920-5000
TS14920-0025EA 217 USD
TS14920-0025 TS14920-2500 TS14920-5000
Methacrylamide 98%, Extra Pure
Methacrylamide
Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 215 °C (760 mmHg)
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00008018
CAS Number: 79-39-0
EINECS: 201-202-3

Order Now

Specification Test Results

Appearance White to light yellow Crystals
Infrared spectrum Conforms
Melting point 108 to 112 °C
Water ≤1 % (K.F.)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR