Glyoxal 40% in aqueous solution

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethandial

15622-5000 15622-0010 15622-0025 15622-0250
TS15622-5000EA 69 USD
TS15622-5000 TS15622-0010 TS15622-0025 TS15622-0250
Glyoxal 40% in aqueous solution
Glyoxal
Formula: C₂H₂O₂
MW: 58.04 g/mol
Melting Pt: –14 °C
Density: 1.265 g/cm³
Flash Pt: >104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006957
CAS Number: 107-22-2
EINECS: 203-474-9

Order Now

Specification Test Results

Appearance Colorless to yellow Solution (May darken on storage)
Titration excess NaOH 39 to 41 %
Acidity (CH3COOH) ≤0.25 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR