D-(-)-Ribose 99+%
Supplier: Thermo Scientific Chemicals
13236-0050
13236-0250
13236-1000
13236-5000
TS13236-0050EA
29
USD
TS13236-0050
TS13236-0250
TS13236-1000
TS13236-5000
D-(-)-Ribose 99+%
D(-)-Ribose
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol Melting Pt: 88…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135453 CAS Number: 50-69-1 EINECS: 200-059-4 Merck Index: 12,08371 |
Specification Test Results
Appearance | White to light yellow to light beige Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % (40°C, 5 h, 20 Torr) |
Sulfated ash | ≤0.1 % |
HPLC | ≥99.0 % (ELSD) |
Specific optical rotation | -19.2° to -20.8° (20°C, 589 nm) (c=4, H2O) |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...