5-Bromouracil 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromopyrimidine-2,4(1H,3H)-dione, Bromouracil

22644-0050 22644-0250
TS22644-0050EA 27 USD
TS22644-0050 TS22644-0250
5-Bromouracil 98%
5-Bromouracil
Formula: C₄H₃BrN₂O₂
MW: 190.98 g/mol
Melting Pt: 307…310 °C
Storage Temperature: Ambient
MDL Number: MFCD00006017
CAS Number: 51-20-7
EINECS: 200-084-0
Merck Index: 13,01425

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR