5-(Chloromethyl)uracil 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-(Chloromethyl)pyrimidine-2,4(1H,3H)-dione

44619-0010 44619-0050
TS44619-0010EA 218 USD
TS44619-0010 TS44619-0050
5-(Chloromethyl)uracil 97%
5-(Chloromethyl)uracil
Formula: C₅H₅ClN₂O₂
MW: 160.56 g/mol
Melting Pt: >350 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00218445
CAS Number: 3590-48-5

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR