2-Thiouracil 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxypyrimidine-2(1H)-thione

15184-0050 15184-1000 15184-5000
TS15184-0050EA 21 USD
TS15184-0050 TS15184-1000 TS15184-5000
2-Thiouracil 99%
2-Thiouracil
Formula: C₄H₄N₂OS
MW: 128.15 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006039
CAS Number: 141-90-2
EINECS: 205-508-8
UN: 3077
Merck Index: 12,09504

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 3 h)
HPLC ≥98.0 %
Appearance of solution (0.5% in 1M NaOH) Clear to light hazy

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR