2-Mercapto-1-methylimidazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12550-0250
12550-1000
12550-5000
TS12550-0250EA
58
USD
TS12550-0250
TS12550-1000
TS12550-5000
2-Mercapto-1-methylimidazole 98%
2-Mercapto-1-methylimidazole
Formula:
C₄H₆N₂S Boiling Pt: 280 °C (760 mmHg) Melting Pt: 140…145 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00179321 CAS Number: 60-56-0 EINECS: 200-482-4 UN: 3335 ADR: 9,III Merck Index: 13,06000 |
Specification Test Results
Appearance | White to cream to yellow-brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 140 to 145 °C |
Loss on drying | ≤1 % (105 °C, 2 hrs) |
Titration with NaOH | ≥97.5 % (On dried substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...