2-Deoxy-D(-)-ribose 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal

15328-0050 15328-0250
TS15328-0050EA 91 USD
TS15328-0050 TS15328-0250
2-Deoxy-D(-)-ribose 99%
2-Deoxy-D(-)-ribose
Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00135904
CAS Number: 533-67-5
EINECS: 208-573-0
Merck Index: 13,02925

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Assay ≥98.5 %
Loss on drying ≤1.0 % (40°C, 2 hrs) (vacuum)
Residue after ignition ≤0.5 %
Specific optical rotation -55° to -59° (20°C, 589 nm) (c=1, H2O, 24 hrs.)
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR