Uracil 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Pyrimidine-2,4(1H,3H)-dione

15730-0025 15730-0250 15730-1000 15730-5000
TS15730-0025EA 1073 USD
TS15730-0025 TS15730-0250 TS15730-1000 TS15730-5000
Uracil 99+%
Uracil
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Boiling Pt: 379
Melting Pt: 194
Storage Temperature: Ambient
MDL Number: MFCD00006016
CAS Number: 66-22-8
EINECS: 200-621-9
Merck Index: 12,09985

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (115°C, 4 h)
Sulfated ash ≤0.3 %
HPLC ≥99.0 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR