5-Fluorouracil 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

22844-0010 22844-0050 22844-0250
TS22844-0010EA 44 USD
TS22844-0010 TS22844-0050 TS22844-0250
5-Fluorouracil 99%
5-Fluorouracil
Formula: C₄H₃FN₂O₂
MW: 130.08 g/mol
Boiling Pt: 190…200 °C (.1 mmHg)
Melting Pt: 278…282 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006018
CAS Number: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,04208

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (80°C, 4 hrs) (in vacuum)
Sulfated ash ≤0.1 %
Titration with NaOH 98.5 to 101.0 % (On dried substance)
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR